ContactUsLinkComponent
pTYB21 Vector, New England Biolabs
  CA101641-054
 :  
undefined
pTYB21 Vector, New England Biolabs
  CA101641-054
 :  N6709S
 :  
  • Description:
    pTYB21 Vector
  • Size:
    10 µg
  • Cat. no.:
    CA101641-054
  • Vector Details:
    pTYB21
pTYB21 is an E
It is an N-terminal fusion vector designed for in-frame insertion of a target gene into the polylinker, downstream of the intein tag (the Sce VMA intein/chitin binding domain, 55 kDa). This allows the N-terminus of the target protein to be fused to the intein tag. The self-cleavage activity of the intein allows the release of the target protein from the chitin-bound intein tag, resulting in a single column purification of the target protein.
coli cloning and expression vector (7514 bp) used in the IMPACT™ Protein Purification System which allows the overexpression of a target protein as a fusion to a self-cleavable affinity tag.