Phi29 DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
phi29 DNA Polymerase is the replicative polymerase from the Bacillus subtilis phage phi29 (Φ29)
This polymerase has exceptional strand displacement and processive synthesis properties. The polymerase has an inherent 3´→5' proofreading exonuclease activity.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together