ContactUsLinkComponent
pCLIPf Vector, New England Biolabs
Catalog # CA101641-588
CAS Number:  
undefined
pCLIPf Vector, New England Biolabs
Catalog # CA101641-588
Supplier Number:  N9215S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    pCLIPf Vector
  • Size:
    20 µg
  • Cat. no.:
    CA101641-588
  • Vector Details:
    pCLIPf
Specifications
About this item
pCLIPf Vector is a mammalian expression plasmid intended for the cloning and stable or transient expression of CLIP-tag® protein fusions in mammalian cells
This plasmid encodes CLIPf, a CLIP-tag protein, which is expressed under control of the CMV promoter. The expression vector has an IRES (internal ribosome entry site) and a neomycin resistance gene downstream of the CLIPf for the efficient selection of stable transfectants. pCLIPf Vector contains two multiple cloning sites to allow cloning of the fusion partner as a fusion to the N- or C-terminus of the CLIPf.