NEBuffer 4 , New England Biolabs
NEBuffer 4, 6.0 mL
Catalog # CA101227-464
Supplier: New England Biolabs (NEB)
Fournisseur #: B7004S
NEBuffer 4 , New England Biolabs
Catalog # CA101227-464
Supplier: New England Biolabs (NEB)
CAS Number:
Jump to:
Product Details & Documents
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity
The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1, NEBuffer 2.1, NEBuffer 3.1 and CutSmart Buffer are available.
Spécifications
Spécifications
- Désignation:NEBuffer 4
- Taille:6.0 mL