NEBNEXT® ULTRA™ DNA Library Prep Kit for Illumina, New England Biolabs
Fournisseur: New England Biolabs (NEB)
Certificats
À propos de cet article
The NEBNext® Ultra™ DNA Library Prep Kit for Illumina contains enzymes and buffers that are ideal to convert a small amount of DNA input into indexed libraries for next-generation sequencing on the Illumina platform (Illumina, Inc)
The workflow of NEBNext Ultra DNA Library Prep Kit for Illumina is very user-friendly and fast with minimal hands-on time. Each of these components must pass rigorous quality control standards and are lot controlled, both individually and as a set of reagents.
Frequently Bought Together