NEBNEXT® QUICK LIGATION™ Reaction Buffer (5X), New England Biolabs
Catalogue # CA101710-268
Fournisseur: New England Biolabs (NEB)
Numéro CAS:
Spécifications
- Description:NEBNext Quick Ligation Reaction Buffer (5X)
- Size:2.0 mL
- Cat. no.:CA101710-268
Spécifications
À propos de cet article
New England Biolabs supplies a 5X reaction buffer for use with Quick T4 DNA Ligase
At a 1X concentration this reaction buffer assures optimal activity of the enzyme.