NEBNEXT® Multiplex Small RNA Library Prep Set for Illumina (1-12), New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
The novel NEBNext® Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries
The NEBNext® Small RNA Library Prep Set for Illumina® (Set 1) includes adaptors and multiplex primers with 12 indices, to enable multiplexing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together