NEBNEXT® mRNA Second Strand Synthesis Module, New England Biolabs
Fournisseur: New England Biolabs (NEB)
Certificats
À propos de cet article
The NEBNext Second Strand Synthesis Module has been optimized to generate double stranded cDNA from first strand cDNA
- NEBNext Second Strand Synthesis (dNTP-free) reaction buffer
The NEBNext Second Strand Synthesis Module is provided as a master mix to maximize efficiency and convenience in RNA sample preparation workflows.
Frequently Bought Together