ContactUsLinkComponent
NEBNEXT® End Repair Reaction Buffer, New England Biolabs
  CA101710-270
 :  
undefined
NEBNEXT® End Repair Reaction Buffer, New England Biolabs
  CA101710-270
 :  B6052S
 :  
  • Description:
    NEBNext End Repair Reaction Buffer
  • Size:
    2.0 mL
  • Cat. no.:
    CA101710-270
New England Biolabs supplies a 10X reaction buffer for use with NEBNext® End Repair Enzyme Mix
At a 1X concentration this reaction buffer assures optimal activity of the enzyme mix.