NEBNEXT® End Repair Module, New England Biolabs
Fournisseur: New England Biolabs (NEB)
Certificats
À propos de cet article
The NEBNext® End Repair Module has been optimized to repair DNA fragmented by nebulization, acoustic shearing, or nucleases
Fragmented DNA is converted by the NEBNext End Repair Module to blunt ended DNA having 5´-phosphates and 3´-hydroxyl. The NEBNext End Repair Module is provided as a master mix to maximize efficiency and convenience in DNA sample preparation workflows.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together