NEBNEXT® dA-Tailing Reaction Buffer, New England Biolabs
Catalogue # CA101710-272
Fournisseur: New England Biolabs (NEB)
Numéro CAS:
Spécifications
- Description:NEBNext dA-Tailing Reaction Buffer
- Size:2.0 mL
- Cat. no.:CA101710-272
Spécifications
À propos de cet article
New England Biolabs supplies a unique 10X reaction buffer for use with NEBNext® Second Strand Synthesis Enzyme Mix when alternative concentrations of deoxynucleotides are required.