Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:NEB 10-beta electrocompetent E.coli
- Size:6×0.1 ml
- Cat. no.:CA101417-922
Specifications
About this item
NEB 10-beta electrocompetent E. coli cells are optimized for high efficiency transformation by electroporation. These cells are ideal for DNA library constructions and all cloning purposes.
- High efficiency strain ideal for cloning large plasmids and BACs
- Also available in chemically-competent format (single-use and 200 µl vials)
- No dry ice surcharge on competent cell shipments
- Outgrowth medium included
- Free of animal products