ContactUsLinkComponent
M13mp18 Single-Stranded DNA, New England Biolabs
Catalog # CA101227-718
CAS Number:  
undefined
M13mp18 Single-Stranded DNA, New England Biolabs
Catalog # CA101227-718
Supplier Number:  N4040S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    M13mp18 Single-stranded DNA
  • Size:
    10 µg
  • Cat. no.:
    CA101227-718
Specifications
About this item
The single-stranded viral DNA is isolated from M13mp18
M13mp18 is a M13 lac phage vector which contains single HindIII, SphI, SbfI, PstI, SalI (AccI/ HincII), XbaI, BamHI, SmaI (XmaI), KpnI (Acc65I), SacI and EcoRI sites within the gene encoding β-Galactosidase.This DNA preparation is useful as a standard and has been tested as a template in the dideoxy-nucleotide termination method of sequencing DNA. M13mp18 phage is propagated in E. coli ER2738. The phage is purified by polyethylene glycol precipitation. The single-stranded DNA is then extracted with phenol.