ContactUsLinkComponent
M13mp18 RF I DNA, New England Biolabs
Catalogue # CA101228-156
Numéro CAS:  
undefined
M13mp18 RF I DNA, New England Biolabs
Catalogue # CA101228-156
Supplier Number:  N4018S
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    M13mp18 RF I DNA
  • Size:
    10 µg
  • Cat. no.:
    CA101228-156
Spécifications
À propos de cet article
M13mp18 is the double-stranded, covalently closed, circular form of DNA derived from bacteriophage M13.
This phage vector contains single HindIII, SphI, SbfI, PstI, SalI (AccI/ HincII), XbaI, BamHI, SmaI (XmaI), KpnI (Acc65I), SacI and EcoRI sites within the β-Galactosidase gene. When a fragment of DNA is inserted into one of these sites, the β-Galactosidase gene is inactivated, providing selection for clones on the appropriate indicator plate. DNASU and Addgene are central repositories for plasmid clones and collections that may also be helpful. The phage M13mp18 is propagated in E. coli ER2738. The replicative form of DNA is isolated from infected cells and purified by a standard plasmid purification procedure. The final preparation is tested for its suitability as a vector.