Lambda DNA, New England Biolabs | Avantor
Lambda DNA, New England Biolabs
Lambda DNA, New England Biolabs
Certificates
Lambda DNA, New England Biolabs
Lambda DNA, New England Biolabs
2 Options Available Below
Product Details & Documents
Duplex DNA is isolated from bacteriophage lambda (cI857ind 1 Sam 7)
Lambda DNA is 48,502 base pairs in length. The phage is isolated from the heat-inducible lysogen E. coli l cI857 S7. The DNA is isolated from the purified phage by phenol extraction and dialyzed against 10 mM Tris-HCl (pH 8.0), 1 mM EDTA.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Lambda DNA
    1,250 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $486.00
    $486.00
    Each
       $486.00
    La quantité minimale de commande est de 1
    Supplier Number: N3011L
  • Lambda DNA
    250 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $124.00
    $124.00
    Each
       $124.00
    La quantité minimale de commande est de 1
    Supplier Number: N3011S