Klenow Fragment (3'-5' exo-)
Catalog # CA101228-220
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Size:1000 units
- Enzyme name:Klenow Fragment (3'-5' exo-)
- Enzyme activity:50000 u/ml
- Cat. no.:CA101228-220
Specifications
About this item
Klenow Fragment (3´→ 5´ exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5´→ 3´ exonuclease activity and has mutations (D355A, E357A) which abolish the 3´→ 5´ exonuclease activity.