Human Alkyladenine Glycosylase (hAAG)
Catalog # CA101449-300
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Species:Human
- Size:500 units
- Enzyme name:Human Alkyladenine Glycosylase
- Enzyme synonyms:hAAG
- Cat. no.:CA101449-300
Specifications
About this item
Human Alkyladenine DNA Glycosylase (hAAG) excises alkylated and oxidative DNA damaged sites, including 3-methyladenine, 7-methylguanine, 1,N6-ethenoadenine and hypoxanthine
hAAG catalyzes the hydrolysis of the N-glycosidic bond to release the damaged base. hAAG is also known as methylpurine DNA glycosylase (MPG) or 3-methyladenine-DNA glycosylase (ANPG).