Exonuclease T
Supplier: New England Biolabs (NEB)
Certificates
About this item
Exonuclease T (Exo T), also known as RNase T, is a single-stranded RNA or DNA specific nuclease that requires a free 3´ terminus and removes nucleotides in the 3´→ 5´ direction
Exonuclease T can be used to generate blunt ends from RNA or DNA molecules that have 3´ extensions.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme name:Exonuclease T