ContactUsLinkComponent
dsRNA Ladder, New England Biolabs
Catalog # CA200064-448
CAS Number:  
undefined
dsRNA Ladder, New England Biolabs
Catalog # CA200064-448
Supplier Number:  N0363S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    dsRNA Ladder
  • Size:
    25 gel lanes
  • Cat. no.:
    CA200064-448
Specifications
About this item
The dsRNA Ladder is a set of 7 annealed double-stranded RNA molecules produced by in vitro transcription of 14 linear DNA templates
The ladder sizes are: 500, 300, 150, 80, 50, 30 and 21 base pairs. All have 2-base, 3´ extensions. The 80 base pair fragment is at double intensity to serve as a reference point. This ladder is suitable for use as a size standard in dsRNA and RNAi analysis on both non-denaturing polyacrylamide and agarose gels.