Deoxynucleotide Solution Set, New England Biolabs
Deoxynucleotide Solution Set, 25 µmol of each
Supplier: New England Biolabs (NEB)
: N0446S
Deoxynucleotide Solution Set, New England Biolabs
Catalog # CA101227-334
Supplier: New England Biolabs (NEB)
CAS Number:
Jump to:
Product Details & Documents
The Deoxynucleotide (dNTP) Solution Set contains 0.25 ml of each solution of ultrapure deoxynucleotide (dATP, dCTP, dGTP and dTTP)
Each deoxynucleotide is supplied at a concentration of 100 mM. Supplied in: Ultrapure water as a sodium salt at pH 7.5.
Spécifications
Spécifications
- Désignation:Deoxynucleotide Solution Set
- Taille:25 µmol of each