DEEP VENT™ (exo-) DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
Deep VentR (exo-) DNA Polymerase has been genetically engineered to eliminate the 3´→ 5´ proofreading exonuclease activity associated with Deep Vent DNA Polymerase
Deep Vent (exo-) DNA Polymerase is even more stable than VentR® (exo-) DNA Polymerase with a half-life of 23 hours at 95°C and 8 hours at 100°C.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together