dATP Solution, New England Biolabs
dATP Solution, 25 µmol
CA101641-036
Supplier #: N0440S
dAtp Solution contains 0.25 ml of a 100 mM solution of ultrapure dATP as a sodium salt at pH 7.4.
- Désignation:dATP Solution
- Taille:25 µmol