About this item
Human apurinic/apyrimidinic (AP) endonuclease, APE 1, also known as HAP 1 or Ref-1, shares homology with E
APE 1 cleaves the phosphodiester backbone immediately 5´ to an AP site via hydrolytic mechanism, generating a ssDNA break that leaves a 3´-hydroxyl and 5´-deoxyribose phosphate terminus. Besides AP endonuclease activity, APE 1 has also been reported to have weak DNA 3´-diesterase, 3´ to 5´ exonuclease and RNase H activities. In addition to DNA repair, APE 1 can also regulate the DNA binding activity of many transcription factors in vitro. APE 1 can also stimulate the DNA binding activity of Fos-Jun heterodimers, Jun-Jun homodimers and HeLa cell AP-1 proteins as well as that of several other transcription factors including NF-kB, Myb and members of the ATF/CREB family.
coli Exonuclease III.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme name:APE 1
- Tested applications:Restriction Enzyme