AMV Reverse Transcriptase, New England Biolabs
Avian Myeloblastosis Virus (AMV) Reverse Transcriptase is an RNA-directed DNA polymerase
This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template.
Frequently Bought Together