ContactUsLinkComponent
Alpha 1-3,6 Galactosidase
Catalogue # CA101447-570
Numéro CAS:  
undefined
Alpha 1-3,6 Galactosidase
Catalogue # CA101447-570
Supplier Number:  P0731S
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    100 units
  • Enzyme name:
    alpha 1-3,6 Galactosidase
  • Cat. no.:
    CA101447-570
Spécifications
À propos de cet article
α1-3, 6 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of α1-3, 6 linked D-galactopyranosyl residues from oligosaccharides.