ContactUsLinkComponent
Magnetic Separation Rack, 50 ml, New England Biolabs
Catalogue # CA200064-490
undefined
Magnetic Separation Rack, 50 ml, New England Biolabs
Catalogue # CA200064-490
Supplier Number:  S1507S
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    50 mL Magnetic Separation Rack
  • Cat. no.:
    CA200064-490
Spécifications
À propos de cet article
The Magnetic Separation Racks are designed to be used for small-scale separations using magnetic particles
The magnets are located on the sides of the racks, resulting in minimal sample loss during pipetting.