WaterPro® Bladder Tanks, Labconco | Avantor
WaterPro® Bladder Tanks, Labconco
Supplier:   Labconco
WaterPro® Bladder Tanks, Labconco
Supplier:  Labconco
Certificates
WaterPro® Bladder Tanks, Labconco
WaterPro® Bladder Tanks, Labconco
2 Options Available Below
Product Details & Documents
WaterPro® RO Bladder Tanks provide additional storage of reverse osmosis (RO) water produced by the WaterPro® RO System (sold separately)
The pressurized water can be distributed to a polishing system or glassware washer through either of two pressurized ports. Includes all tubing and hardware for connection. Available in 14 or 20 gallon sizes.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • WaterPro® RO Bladder Tank, 14 Gal. (53 L), for WaterPro® RO System
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,480.00
    $2,480.00
    Each
       $2,480.00
    La quantité minimale de commande est de 1
    Supplier Number: 9201001
  • WaterPro® RO Bladder Tank, 20 Gal. (75.7 L), for WaterPro® RO System
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,730.00
    $2,730.00
    Each
       $2,730.00
    La quantité minimale de commande est de 1
    Supplier Number: 9201000