Vacuum Adapter, with Outer Joint, Glassco
Supplier: Glassco
Jump to:
À propos de cet article
This vacuum adapter features an outer standard taper joint.
- Top outer [ST] joint
- Manufactured from borosilicate 3.3 glass
Adapter is ideal for use as a vent, or for the introduction of pressure or vacuum.
1 Options Available Below
Product Details & Documents
This vacuum adapter features an outer standard taper joint.
- Top outer [ST] joint
- Manufactured from borosilicate 3.3 glass
Adapter is ideal for use as a vent, or for the introduction of pressure or vacuum.
Product Family Options
Informations sur le produit
- DésignationTop ST Joint SizeAvailabilityPrice
- Supplier Number: 020.470.03
- Discontinued-Supplier Number: -