Reflectoquant® pH Test, MilliporeSigma
Reflectoquant® pH Test, MilliporeSigma
Reflectoquant® pH Test, MilliporeSigma
Supplier:  MilliporeSigma
Certificates
Reflectoquant® pH Test, MilliporeSigma
1 Options Available Below
Product Details & Documents
In water analysis the pH value plays a very important role in how other substances behave.
The test can be used for monitoring the pH value in aqueous solutions, beverages, e.g. wine, after dilution and food after sample pretreatment.
Product Family Options
Product Information
  • Désignation
    Taille
    pH Range
    Availability
    Price
  • pH test method, reflectometric with test strips
    50 strips
    1.0 - 5.0
    Discontinued
    Supplier Number: -
  • pH test method, reflectometric with test strips
    50 strips
    4.0 - 9.0
    Log in for availability
    Log in for availability
    Each
    Each - $84.83
    $84.83
    Each
       $84.83
    Supplier Number: 1.16996.0001