Peroxide Test, MQuant®, MilliporeSigma
Peroxide Test, MQuant®, MilliporeSigma
Peroxide Test, MQuant®, MilliporeSigma
The peroxide test is ideal for testing residual concentrations of disinfectants on equipment or the product itself following cleaning and rinsing.
Product Family Options
  • Désignation
    Méthode chimique
    Range
    No. of Tests
  • Test with Strips
    Enzymatic reaction
    0,5-2-5-10-25 mg/l H₂O₂
    25 strips
    Each
    Each - $96.47
    $96.47
    Each
       $96.47
     : 1.10011.0002
  • Test with Strips
    Enzymatic reaction
    0,5-2-5-10-25 mg/l H₂O₂
    100 strips
    Each
    Each - $185.89
    $185.89
    Each
       $185.89
     : 1.10011.0001