Lab Armor® Walkabout Trays, Shel Lab
The Walkabout Tray is jaw-droppingly clever
The Walkabout Tray can maintain a 5 ml liquid sample at 37°C ± 1.3°C for 30 minutes after removing it from the water bath. It is durable and lightweight, perfect for use under the hood. Molded one-piece construction, condensation free.
Does not include a lid. Hand wash with mild detergent, towel dry.
It's a quarter the size of a traditional lab bucket. Making it easy to use under the hood or in tight benchtop spaces. With its superior insulative properties, it keeps your samples and reagents at temperature after removing them from the bead bath, incubator or refrigerator.
- Catalog No:
- 89409-260
- 89409-258
- Capacity:
- 0.5 – 1.0 L (beads)
- 0.5 – 1.0 L (beads)
- Temperature range:
- -196 to 93°C
- -196 to 93°C
- Dimensions:
- 15.24 x 15.24 x 7.6 cm (6 x 6 x 3")
- 15.24 x 15.24 x 7.6 cm (6 x 6 x 3")
Frequently Bought Together