Specifications
- Temperature Range:5 °C above ambient...80 °C
- Capacity:20 L (tank)
- Description:Bead Bath
- Exterior dimensions:21.6 x 40.4 x 59 cm (8.5 x 15.9 x 23.25")
- Dimensions:15.2 x 30.5 x 43.2 cm (6 x 12 x 17"), tub
- Cat. no.:89409-224
- Electrical:100 - 120 V, 50/60 Hz, 6 A
- Uniformity:±1.0 °C
- Supplier No.:74300-720
Specifications
About this item
The Bead Bath's eco-friendly, state-of-the-art design takes full advantage of the robust properties of Lab Armor Beads
Lab Armor Digital Baths are durable, dependable, and are available with a variety of safety and convenience features: Digital microprocessor control, Over-temperature protection, easy-to-clean stainless steel body and lid, surround heating eliminates hot spots, compatible with beads only.
Eco-Friendly. Eliminates harmful biocides, Uses Lab Armor Beads, made from solid recyclable metal. More energy efficient, uses over 4X less energy when set to 65°C and over 2X less energy at 37°C.
It delivers exceptional temperature uniformity and gets up and running faster. So you can do things with this bath that you can't do with your old water bath. You aren't limited to water tight containers, so you can safely incubate multi-well plates, petri dishes, and open-top samples at any angle.