ContactUsLinkComponent
Capto™ Q Ion Exchange Chromatography Media
Capto™ Q Ion Exchange Chromatography Media
Supplier:  Cytiva
Certificates
undefined
Capto™ Q Ion Exchange Chromatography Media
Supplier:  Cytiva
Certificates
Specifications
  • pH range:
    2 - 12
  • Matrix:
    Highly cross-linked agarose with dextran surface extender
  • Ligand:
    Quaternary amine
  • Storage temperature:
    4 - 30 °C
  • Total ionic capacity:
    0,16 - 0,22 mmol Cl-/ml