ContactUsLinkComponent
rProtein A Sepharose™ Fast Flow Affinity Purification Media, Cytiva
Fournisseur:  Cytiva
Certificats
undefined
rProtein A Sepharose™ Fast Flow Affinity Purification Media, Cytiva
Fournisseur:  Cytiva
Certificats
Spécifications
  • pH range:
    3 - 10
  • Matrix:
    Cross-linked agarose, 4%, spherical
  • Ligand:
    Recombinant protein A (E.coli)
  • Storage temperature:
    2 - 8 °C
  • Binding capacity:
    ~50 mg hIgG/ml
  • Ligand density:
    6 mg rProtein A/ml
  • Ligand coupling method:
    Epoxy activation
  • Flow rate:
    150 - 250 cm/h