Ab Buffer Kit, Cytiva
Ab Buffer Kit
Catalog # CA95056-236L
Supplier:   Cytiva
 :  28-9030-59
Ab Buffer Kit, Cytiva
Catalog # CA95056-236L
Supplier:  Cytiva
CAS Number:  
Ab Buffer Kit, Cytiva
Ab Buffer Kit, Cytiva
Ab Buffer Kit
CAS Number:  
Price
$466.78
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Ab Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of IgG according to recommended protocols.
The kit also includes neutralizing buffer. Ab Buffer Kit eliminates time consuming buffer preparation, which allows fast, reproducible, and convenient purification work.
 : Includes 50 ml binding buffer (10X stock solution), 15 ml elution buffer (10X stock solution), and 25 ml neutralizing buffer.
Specifications
Specifications
  • Désignation:
    Ab Buffer Kit
  • Cat. No.:
    CA95056-236L
  • Storage temperature:
    4–8°C, 20% Ethanol
Specifications