À propos de cet article
Absorbs the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant pillow won't degrade or cause a dangerous reaction upon contact with corrosive spills. Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids; great for adding absorbency during spill response. Polypropylene skin resists chemicals and tearing; reduces dust and holds in liquid, even when saturated. Polypropylene filler is highly absorbent for containing corrosive or reactive spills.
- Made of 100% chemical-resistant skin and fast-wicking filler
- Large surface area and high capacity to quickly soak up liquids
Absorbs and retains most acids, bases and unknown liquids. Pink color is easily distinguishable to assure workers use the correct pillow during a spill emergency. Can be incinerated after use to reduce waste.