Syringe Filters, Argos Technologies | Avantor
Syringe Filters, Argos Technologies
Syringe Filters, Argos Technologies
Supplier:  Antyila Scientific
Certificates
Syringe Filters, Argos Technologies
Syringe Filters, Argos Technologies
17 Options Available Below
Product Details & Documents
Color coded for easy membrane identification.
  • Available in 13 and 30 mm diameters
  • Choice of 0.45 and 0.22 µm membranes
  • Designed for low hold-up to minimize sample loss
  • Lot numbers are printed on sterile packs for easy traceability
  • Sterile filters are packaged in a convenient, peel-open package
  • All products come certified non-pyrogenic
Syringe Filters are ideal for filtering culture media and organic solvents in a wide range of applications.
New surfactant-free cellulose acetate filter membranes contain none of the wetting agents found in standard CA (cellulose acetate). These wetting agents have been shown to be toxic to specific cell lines in vitro (1 - 3). SFCA also has lower protein binding than does standard CA.
SFCA filters feature:
Low protein binding with little or no loss of specific proteins
Excellent flow rates
No wetting agents to affect sensitive cell culture lines
Much lower extractables than standard CA membranes
Membrane material includes SFCA, MCE, PVDF, PTFE, Nylon, and PES. All items are tested for filter efficiency and housing integrity. Filter housing is manufactured and tested to withstand pressures of up to 75 psi (5.1 bar). Filters are available nonsterile or sterile, individually wrapped (sterile), certified non-pyrogenic and RNase-/DNase-free.
Documents
Product Family Options
Surfactant-Free Cellulose Acetate Membrane
Informations sur le produit
  • Désignation
    Ø
    Taille de pore
    Color
    Sterility
    Packaging
    Availability
    Price
  • Syringe Filters
    30 mm
    0.22 µm
    Red
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $1,180.31
    $1,180.31
    Prix promotionnel pour les nouveaux clients
    Pack of 45
       $1,180.31
    Supplier Number: 04395-90
  • Syringe Filters
    30 mm
    0.22 µm
    Red
    Non sterile
    Bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $601.53
    $601.53
    Prix promotionnel pour les nouveaux clients
    Pack of 50
       $601.53
    Supplier Number: 04395-92
  • Syringe Filters
    30 mm
    0.45 µm
    Red
    Non sterile
    Bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $819.81
    $819.81
    Prix promotionnel pour les nouveaux clients
    Pack of 50
       $819.81
    Supplier Number: 04395-93
MCE Membrane
Informations sur le produit
  • Désignation
    Ø
    Taille de pore
    Color
    Sterility
    Packaging
    Availability
    Price
  • Syringe Filters
    30 mm
    0.22 µm
    Yellow
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $626.71
    $626.71
    Prix promotionnel pour les nouveaux clients
    Pack of 45
       $626.71
    La quantité minimale de commande est de 1
    Supplier Number: 04396-20
  • Syringe Filters
    30 mm
    0.45 µm
    Yellow
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $611.39
    $611.39
    Prix promotionnel pour les nouveaux clients
    Pack of 45
       $611.39
    La quantité minimale de commande est de 1
    Supplier Number: 04396-21
Nylon Membrane
Informations sur le produit
  • Désignation
    Ø
    Taille de pore
    Color
    Sterility
    Packaging
    Availability
    Price
  • Syringe Filters
    13 mm
    0.45 µm
    Pink
    Non sterile
    Bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $579.98
    $579.98
    Prix promotionnel pour les nouveaux clients
    Pack of 100
       $579.98
    La quantité minimale de commande est de 1
    Supplier Number: 04396-22
  • Syringe Filters
    30 mm
    0.45 µm
    Pink
    Non sterile
    Bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $694.57
    $694.57
    Prix promotionnel pour les nouveaux clients
    Pack of 50
       $694.57
    La quantité minimale de commande est de 1
    Supplier Number: 04396-23
  • Syringe Filters
    30 mm
    0.22 µm
    Pink
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $890.63
    $890.63
    Prix promotionnel pour les nouveaux clients
    Pack of 45
       $890.63
    La quantité minimale de commande est de 1
    Supplier Number: 04396-24
  • Syringe Filters
    30 mm
    0.45 µm
    Pink
    Sterile
    Individually Wrapped
    Discontinued
    Supplier Number: 04396-25
PES Membrane
Informations sur le produit
  • Désignation
    Ø
    Taille de pore
    Color
    Sterility
    Packaging
    Availability
    Price
  • Syringe Filters
    13 mm
    0.22 µm
    Green
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $1,109.10
    $1,109.10
    Prix promotionnel pour les nouveaux clients
    Pack of 100
       $1,109.10
    La quantité minimale de commande est de 1
    Supplier Number: 04396-26
  • Syringe Filters
    13 mm
    0.45 µm
    Green
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $1,136.43
    $1,136.43
    Prix promotionnel pour les nouveaux clients
    Pack of 100
       $1,136.43
    La quantité minimale de commande est de 1
    Supplier Number: 04396-27
  • Syringe Filters
    30 mm
    0.22 µm
    Green
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $1,216.83
    $1,216.83
    Prix promotionnel pour les nouveaux clients
    Pack of 45
       $1,216.83
    La quantité minimale de commande est de 1
    Supplier Number: 04396-28
  • Syringe Filters
    30 mm
    0.45 µm
    Green
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $1,709.88
    $1,709.88
    Prix promotionnel pour les nouveaux clients
    Pack of 45
       $1,709.88
    La quantité minimale de commande est de 1
    Supplier Number: 04396-29
PTFE Membrane
Informations sur le produit
  • Désignation
    Ø
    Taille de pore
    Color
    Sterility
    Packaging
    Availability
    Price
  • Syringe Filters
    30 mm
    0.22 µm
    White
    Non sterile
    Bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $910.01
    $910.01
    Prix promotionnel pour les nouveaux clients
    Pack of 50
       $910.01
    La quantité minimale de commande est de 1
    Supplier Number: 04396-30
  • Syringe Filters
    30 mm
    0.45 µm
    White
    Non sterile
    Bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $623.92
    $623.92
    Prix promotionnel pour les nouveaux clients
    Pack of 50
       $623.92
    La quantité minimale de commande est de 1
    Supplier Number: 04396-31
PVDF Membrane
Informations sur le produit
  • Désignation
    Ø
    Taille de pore
    Color
    Sterility
    Packaging
    Availability
    Price
  • Syringe Filters
    13 mm
    0.22 µm
    Blue
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $1,244.88
    $1,244.88
    Prix promotionnel pour les nouveaux clients
    Pack of 100
       $1,244.88
    La quantité minimale de commande est de 1
    Supplier Number: 04396-32
  • Syringe Filters
    30 mm
    0.22 µm
    Blue
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $1,284.14
    $1,284.14
    Prix promotionnel pour les nouveaux clients
    Pack of 45
       $1,284.14
    La quantité minimale de commande est de 1
    Supplier Number: 04396-33
  • Syringe Filters
    30 mm
    0.45 µm
    Blue
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $1,290.20
    $1,290.20
    Prix promotionnel pour les nouveaux clients
    Pack of 45
       $1,290.20
    La quantité minimale de commande est de 1
    Supplier Number: 04396-34