Accessories for PIG® HazMat Spill Kit in Large Wall-Mount Cabinet, New Pig
Catalog # 56620-642
Supplier: New Pig
Specifications
- Description:Refill for PIG® HazMat Spill Kit in Large Wall-Mount Cabinet
- Cat. no.:56620-642
- Absorbency:68.1 L (18 gal.)
- Supplier No.:RFL328
- Details:80× - 10" W x 13" L PIG® HazMat Chemical Absorbent Handy Pad in Dispenser Box; 3× - ext. dia. 3" x 10' L PIG® HazMat Chemical Absorbent Sock; 2× - ext. dia. 3" x 46" L PIG® HazMat Chemical Absorbent Sock; 12× - 8" W x 8" L x 1" H PIG® HazMat Chemical Absorbent Pillow in Dispenser Box; 40× - 12.25" W x 13" L Polypropylene Wipers; 10× - 18" W x 30" H Polyethylene Disposal Bags; 1× - Instructions
Specifications
About this item
Refill for PIG® HazMat Spill Kit in Large Wall-Mount Cabinet. Don't be caught with an empty spill kit when you need it most! Keep a refill handy to instantly replace used contents.
- Packed to refill your PIG HazMat Spill Kit in Storage Chest or Large Wall-Mount Cabinet
- PIG HazMat Socks stop spreading spills; PIG HazMat Pads and Pillows absorb; wipers clean surfaces and hands; PIG Repair Putty plugs and patches cracks and leaks
- PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
- Temporary disposal bags help make cleanup easier
Only PIG Spill Kit Refills feature PIG Absorbents proven for rapid response.