SP Bel-Art Cleanware™ Label Remover, Bel-Art Products, a part of SP
Catalog # 62866-188
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Label-off™ label remover
- Size:59 ml
- Cat. no.:62866-188
- Supplier No.:170770000
Specifications
About this item
Cleanware™ Label-Off™ Label Remover quickly removes labels, including the gummy residue other cleaners leave behind.
- Quick and easy removal of labels
- No residue left behind
The kit contains 59 ml (2oz.) of label adhesive solvent in a pump spray with cap, along with 20 lint-free, 10.2 cm (4") square wipes.
Ordering information: No air shipment.