BRAND macro™ Pipette Controller, BrandTech
À propos de cet article
macro™ pipette controller provides extremely smooth control.
- Improved design now enables even finer control
- Suitable for volumetric and serological pipettes from 0.1 to 200 ml
The entire macro™ pipette controller can be autoclaved at 121 °C (250 °F) for decontamination. The entire instrument can be disassembled for cleaning and the replacement of soiled filters or other parts.
Simply squeeze the suction bellows, which is sufficient for volumes up to 50 ml. Deflect the pipetting lever upward to aspirate, or downward to dispense. The further the deflection, the faster the aspiration and dispensing speeds. A new spring loaded pipetting lever allows even greater control with small volume pipettes. The proprietary 3 µm filter allows proper drain times for pipetting of Class A glassware, and helps prevent aqueous liquid penetration. A convenient blow out bulb allows accurate use of 'To Contain' pipettes. The suction bellows has been redesigned for even easier operation.
Ordering information: Each macro™ pipette controller is supplied in recyclable packaging with Operating Manual and spare 3 µm filter.
4 Options Available Below
Product Details & Documents
macro™ pipette controller provides extremely smooth control.
- Improved design now enables even finer control
- Suitable for volumetric and serological pipettes from 0.1 to 200 ml
The entire macro™ pipette controller can be autoclaved at 121 °C (250 °F) for decontamination. The entire instrument can be disassembled for cleaning and the replacement of soiled filters or other parts.
Simply squeeze the suction bellows, which is sufficient for volumes up to 50 ml. Deflect the pipetting lever upward to aspirate, or downward to dispense. The further the deflection, the faster the aspiration and dispensing speeds. A new spring loaded pipetting lever allows even greater control with small volume pipettes. The proprietary 3 µm filter allows proper drain times for pipetting of Class A glassware, and helps prevent aqueous liquid penetration. A convenient blow out bulb allows accurate use of 'To Contain' pipettes. The suction bellows has been redesigned for even easier operation.
Ordering information: Each macro™ pipette controller is supplied in recyclable packaging with Operating Manual and spare 3 µm filter.
Product Family Options
Informations sur le produit
- DésignationCapacitéColorAvailabilityPrice
- Supplier Number: 26200
- Supplier Number: 26202
- Supplier Number: 26203
- Supplier Number: 26201
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 3 µm PTFE filter with PP housing, pack of 10Replacement 3 µm PTFE filter with PP housing, pack of 1018900-012Log in pour la disponibilitéPack of 10 - $338.84
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement adapter housing for macro™
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement valve system for macro™, PP/siliconeReplacement valve system for macro™, PP/silicone18900-288Log in pour la disponibilitéEach - $145.89
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement silicone adapter for macro™
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 3 µm PTFE filter with PP housing, individually wrappedReplacement 3 µm PTFE filter with PP housing, individually wrapped18900-282Log in pour la disponibilitéEach - $49.84
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement suction bellows for macro™, gray