Sawtooth Dispersing Tool for T10 Basic Homogenizer, IKA® Works
Fournisseur: IKA Works
Certificats
À propos de cet article
Dispersing tool for T10 Basic homogenizer
Sawtooth design creates a cutting and tearing action for shredding fibrous materials.
Made of high quality stainless steel.
Recommended for use with tissue and other fibrous materials.
Spécifications
- Catalog No:
- 89414-946
- 89414-944
- Gap between rotor and stator:
- 0.25 mm
- 0.25 mm
- Stator Ø:
- 10 mm
- 8 mm
- Rotor Ø:
- 7.5 mm
- 6.1 mm
- Materials in contact with medium:
- PTFE, AISI 316L
- PTFE, AISI 316L
- Max. operating temperature:
- 180°C
- 180°C