Spill-X® Spill Treatment Kit, Acids, New Pig
Catalog # 89205-852
Supplier: New Pig
Specifications
- Description:Shaker Bottles, 6 x 2 lbs.
- Cat. no.:89205-852
- Supplier No.:GEN528
Specifications
About this item
Packed with spill control agents that neutralize and solidify acid spills
Works on most mineral and organic acids for safe and easy cleanup, including sulfuric, hydrochloric, nitric, and hydrofluoric. Granular base lets you use only what you need; reduces product waste and disposal costs. Speeds spill cleanup to limit worker exposure and minimize production downtime. Non-toxic, free-flowing dry chemical is environmentally safe. Agents packed in easy-to-use shaker bottles. Excellent as refill for Spill-X Portable Spill Treatment Kit