MRO Universal Mat Pads, New Pig
Fournisseur: New Pig
Certificats
À propos de cet article
100% polypropylene absorbs water-based liquids, petroleum products and solvents. Dimpled, 3-layer design quickly absorbs liquids. Perforations let you tear off only what you need; reduces waste to save on disposal costs.
- For smaller leak-and-drip applications
- Perforated
Pads are great for spill cleanup and smaller leak-and-drip applications. Rolls are great for catching overspray and machine leaks.
Frequently Bought Together