PIG® Plug-N-Seal® Patching Paste, New Pig
Patching Paste for Fuel Tanks, Steel Drums, and Containers, 1 lb. Container
Catalog # 89205-886
Supplier: New Pig
Fournisseur #: PTY220
Jump to:
Product Details & Documents
Temporarily stops leaks until system can be drained or leak source contained
No preparation needed, even on dirty or rusty surfaces. Temporarily stops leaks from 2 minutes up to 2 hours. Ready to use without mixing.
Excellent chemical resistance makes it excellent for flammable or hazardous leaks.
Spécifications
- Désignation:Patching Paste for Fuel Tanks, Steel Drums, and Containers, 1 lb. Container