PIG® Heavy Fluids Absorbent Mat Roll, New Pig
Catalog # 56620-558
Supplier: New Pig
Specifications
- Description:Heavyweight
- Dimensions:55.9 cm x 26 m (22" x 85')
- Cat. no.:56620-558
- Supplier No.:MAT210
Specifications
About this item
Thirsty air pockets absorb even the stickiest heavy liquids with only slight pressure
Excellent for cleaning up paint spills, glues, resins, liquid chocolate, syrup and varnish. Quickly absorbs thick, high-viscosity liquids that conventional absorbents can't. Can also be used to filter metal chips and other large debris from liquids. Rolls are excellent for cleaning up larger spills of thick liquids.