CRITERION™ Tryptose, Hardy Diagnostics | Avantor
CRITERION™ Tryptose, Hardy Diagnostics
CRITERION™ Tryptose, Hardy Diagnostics
Certificates
CRITERION™ Tryptose, Hardy Diagnostics
2 Options Available Below
Product Details & Documents
CRITERION™ Tryptose is primarily used for preparing microbiological culture media and when non-animal source nutrients are needed in the cultivation of fastidious and non-fastidious organsims
  • Non-mammalian
  • Wide mouth opening
  • Reduces dust and allows easy scooping
  • Convenient handgrip
  • 'No slip' finger holds
  • Stackable containers
  • High solubility; dissolves quickly
  • Custom and bulk products available
Dehydrated culture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: TSE/BSE Free
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • CA89406-666
  • CA89406-668
  • CA89406-672
  • Storage temperature:
  • –2-30°C
  • –2-30°C
  • –2-30°C
Spécifications
Product Family Options
Informations sur le produit
  • Taille
    Final Volume
    Type de conditionnement
    Availability
    Price
  • 122.4 g
    2 L
    Zip-pouch to make 2 L, Mylar®
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89406-666
    Storage temperature
    –2-30°C
  • 500 g
    Bottle, wide mouth
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $265.00
    $265.00
    Each
       $265.00
    La quantité minimale de commande est de 1
    Supplier Number: C8071
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89406-668
    Storage temperature
    –2-30°C
  • 10 kg
    Bucket
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,270.00
    $4,270.00
    Each
       $4,270.00
    La quantité minimale de commande est de 1
    Supplier Number: C8073
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89406-672
    Storage temperature
    –2-30°C