Tryptic Soy Broth (TSB) without Dextrose, CRITERION™, Hardy Diagnostics
Catalog # CA89406-694
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Bottle, wide mouth
- Size:500 g
- Storage temperature:–2-30°C
- Cat. no.:CA89406-694
- Supplier No.:C8121
Specifications
About this item
CRITERION™ Tryptic Soy Broth (TSB) without Dextrose is for the cultivation of microorganisms.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
- BSE Free Documentation Available
- Over 30 years of formulation experience
Certifications: BSE Free