Tryptic Soy Broth (TSB), CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of microorganisms.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Certifications: BSE Free
Specifications
- Storage temperature:–2-30°C