Tryptic Soy Agar (TSA) Blood Base, pH 7.4, CRITERION™, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
À propos de cet article
CRITERION™ TSA (Tryptic Soy Agar) Blood Base, pH 7.4 is for the cultivation of microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
CRITERION™ Dehydrated Culture Media and container.
Certifications: BSE Free
Spécifications
- Storage temperature:–2-30°C