Tryptic Soy Agar (TSA) with Lecithin and Tween, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ Tryptic Soy Agar with Lecithin and Tween® 80 is recommended for use in the detection and enumeration of microorganisms in replicate plating techniques
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Also used for the detection and enumeration of microorganisms present on the surfaces of sanitary importance. CRITERION™ Dehydrated Culture Media and container.
Certifications: BSE Free
Specifications
- Storage temperature:2-8°C